missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Benzothiazolethiol, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 2-Mercaptobenzothiazole |
| CAS | 149-30-4 |
| Molecular Formula | C7H5NS2 |
| Quantity | 100 mg |
| Synonym | 2(3H)-Benzothiazolethione, 2-Benzothiazolethiol (8CI), 2-Benzothiazolinethione (6CI,7CI), 1,3-Benzothiazole-2-thiol, 1,3-Benzothiazole-2-thione, 2,3-Dihydrobenzothiazole-2-thione, 2-Benzothiazolyl mercaptan, 2-MBT, 2-Mercapto-1H-benzothiazole, 2-Mercaptobenzothiazole, 2-Mercaptobenzthiazole, 2-Sulfanylbenzothiazole, Accel M, Accelerator M, Aero 407, Aero Promoter 412, Benz-1,3-thiazolidine-2-thione, Benzo[d]thiazole-2-thiol, Benzothiazolethiol, Captax, Captax MBT, DT 402, Dermacid, Ekagom G, Kaptaks, Kaptax, M 2, M 2 (rust inhibitor), MBT, MBT (vulcanization accelerator), Mebetizol, Mebetizole, Mebithizol, Mercaptobenzothiazole, Mercaptobenzthiazole, Mertax, NSC 2041, Nocceler M, Nocceler M-P, Nonflex NB, Nuodeb 84, OCID, Oricel M, Perkacit MBT, Pneumax MBT, RU 3, Rotax, Royal MBT, Sanceler M, Sanceler M-G, Soxinol M, Thiotax, Vulkacit M, Vulkacit Mercapto, Vulkacit Mercapto MG/C, Vulkacit Mercapto/C, Vulkacit Mercapto/MG, Vulkafil ZN 94TT01, Wobezit M, accelerator MBT |
| InChI Formula | InChI=1S/C7H5NS2/c9-7-8-5-3-1-2-4-6(5)10-7/h1-4H,(H,8,9) |
| SMILES | Sc1nc2ccccc2s1 |
| IUPAC Name | 1,3-benzothiazole-2-thiol |
| Molecular Weight (g/mol) | 167.2513 |
| Formula Weight | 166.9863 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction