missing translation for 'onlineSavingsMsg'
Learn More

2-Benzothiazoleacetonitrile, 98%

Product Code. 11403764
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403764 5 g 5g
11413764 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11403764 Supplier Thermo Scientific Alfa Aesar Supplier No. A11343.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56278-50-3 | C9H6N2S | 174.221 g/mol

2-Benzothiazoleacetonitrile is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dye stuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Benzothiazoleacetonitrile is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dye stuffs.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents .
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56278-50-3
Molecular Formula C9H6N2S
Molecular Weight (g/mol) 174.221
MDL Number MFCD00051633
InChI Key ZMZSYUSDGRJZNT-UHFFFAOYSA-N
Synonym benzothiazole-2-acetonitrile, 2-benzo d thiazol-2-yl acetonitrile, 2-benzothiazoleacetonitrile, 2-1,3-benzothiazol-2-yl acetonitrile, benzothiazole-2-yl-acetonitrile, 1,3-benzothiazol-2-ylacetonitrile, benzothiazol-2-ylacetonitrile, 2-cyanomethyl-1,3-benzothiazole, 2-benzothiazol-2-ylethanenitrile, 2-cyanomethylbenzothiazole
PubChem CID 342465
IUPAC Name 2-(1,3-benzothiazol-2-yl)acetonitrile
SMILES C1=CC=C2C(=C1)N=C(S2)CC#N

Specifications

Melting Point 99°C to 103°C
Quantity 5 g
UN Number UN3439
Beilstein 128460
Solubility Information Slightly soluble in water.
Formula Weight 174.23
Percent Purity 98%
Chemical Name or Material 2-Benzothiazoleacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.