missing translation for 'onlineSavingsMsg'
Learn More

2-Benzo[b]furan-3-ylethanol, ≥95%, Thermo Scientific™

Product Code. 10082014
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10082014 250 mg Amber Glass Bottle 250mg
10677854 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10082014 Supplier Thermo Scientific Maybridge Supplier No. CC40809CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 75611-06-2
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD08271904
InChI Key QBEGKDXITSPUTE-UHFFFAOYSA-N
Synonym 2-benzofuran-3-yl ethanol, 2-benzo b furan-3-ylethanol, 2-1-benzofuran-3-yl ethanol, 2-1-benzofuran-3-yl ethan-1-ol, 2-benzofuran-3-yl ;ethanol, 2-benzo b furan-3-yl ethanol, 2-benzo b furan-3-ylethan-1-ol
PubChem CID 10953836
IUPAC Name 2-(1-benzofuran-3-yl)ethanol
SMILES C1=CC=C2C(=C1)C(=CO2)CCO

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 162.19
Percent Purity 95%
Chemical Name or Material 2-Benzo[b]furan-3-ylethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.