missing translation for 'onlineSavingsMsg'
Learn More

2-Benzo[b]furan-3-ylethanol, ≥95%, Thermo Scientific™

Catalog Number p-3174738
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
75611-06-2
C10H10O2
162.188
MFCD08271904
QBEGKDXITSPUTE-UHFFFAOYSA-N
2-benzofuran-3-yl ethanol, 2-benzo b furan-3-ylethanol, 2-1-benzofuran-3-yl ethanol, 2-1-benzofuran-3-yl ethan-1-ol, 2-benzofuran-3-yl ;ethanol, 2-benzo b furan-3-yl ethanol, 2-benzo b furan-3-ylethan-1-ol
10953836
2-(1-benzofuran-3-yl)ethanol
C1=CC=C2C(=C1)C(=CO2)CCO
This item is not returnable. View return policy
75611-06-2
C10H10O2
162.188
MFCD08271904
QBEGKDXITSPUTE-UHFFFAOYSA-N
2-benzofuran-3-yl ethanol, 2-benzo b furan-3-ylethanol, 2-1-benzofuran-3-yl ethanol, 2-1-benzofuran-3-yl ethan-1-ol, 2-benzofuran-3-yl ;ethanol, 2-benzo b furan-3-yl ethanol, 2-benzo b furan-3-ylethan-1-ol
10953836
2-(1-benzofuran-3-yl)ethanol
C1=CC=C2C(=C1)C(=CO2)CCO
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 162.19
Percent Purity 95%
Chemical Name or Material 2-Benzo[b]furan-3-ylethanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'