missing translation for 'onlineSavingsMsg'
Learn More

2-Benzimidazolepropionic acid, 97%, Thermo Scientific™

Product Code. p-3790860 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10555914

Brand: Thermo Scientific Acros 397780010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 23249-97-0
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.20
MDL Number MFCD00041215
InChI Key XYWJNTOURDMTPI-UHFFFAOYSA-N
Synonym procodazole, 2-benzimidazolepropionic acid, procodazol, 1h-benzimidazole-2-propanoic acid, propazol, 3-1h-benzimidazol-2-yl propanoic acid, procodazole inn, propazole, 3-1h-1,3-benzodiazol-2-yl propanoic acid, unii-sg5iu7fd3r
PubChem CID 65708
IUPAC Name 3-(1H-benzimidazol-2-yl)propanoic acid
SMILES OC(=O)CCC1=NC2=CC=CC=C2N1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 229°C to 231°C
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 7880
Formula Weight 190.2
Percent Purity 97%
Chemical Name or Material 2-Benzimidazolepropionic acid
compliance-icons
Product Identifier
  • 2-Benzimidazolepropionic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.