missing translation for 'onlineSavingsMsg'
Learn More

2-Benzimidazoleacetonitrile, 99%

Product Code. 11344314
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
This item is not returnable. View return policy

Product Code. 11344314

Brand: Thermo Scientific Alfa Aesar L00827.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4414-88-4 | C9H7N3 | 157.176 g/mol

2-Benzimidazoleacetonitrile, is used as a pharmaceutical intermediate. It is also used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Benzimidazoleacetonitrile, is used as a pharmaceutical intermediate. It is also used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4414-88-4
Molecular Formula C9H7N3
Molecular Weight (g/mol) 157.176
MDL Number MFCD00005601
InChI Key BWOVACANEIVHST-UHFFFAOYSA-N
Synonym 2-benzimidazolyl acetonitrile, 2-cyanomethyl benzimidazole, 1h-benzimidazole-2-acetonitrile, 2-benzimidazoleacetonitrile, 2-benzimidazolylacetonitrile, 2-cyanomethylbenzimidazole, 1h-benzimidazol-2-ylacetonitrile, 2-kyanmethylbenzimidazol, benzimidazole, 2-cyanomethyl, 2-1h-1,3-benzodiazol-2-yl acetonitrile
PubChem CID 20455
IUPAC Name 2-(1H-benzimidazol-2-yl)acetonitrile
SMILES C1=CC=C2C(=C1)NC(=N2)CC#N

Specifications

Melting Point 206°C to 211°C
Odor Odorless
Quantity 50 g
Beilstein 128461
Solubility Information Insoluble in water.
Formula Weight 157.18
Percent Purity 99%
Chemical Name or Material 2-Benzimidazoleacetonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.