missing translation for 'onlineSavingsMsg'
Learn More

(-)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%, > 95% ee, Thermo Scientific Chemicals

Product Code. 10155460 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10155460 1 g Glass bottle 1g
10530204 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10155460 Supplier Thermo Scientific Acros Supplier No. 297040010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79200-56-9
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00211274
InChI Key DDUFYKNOXPZZIW-UHNVWZDZSA-N
Synonym 1r,4s-2-azabicyclo 2.2.1 hept-5-en-3-one, --1r,4s-2-azabicyclo 2.2.1 hept-5-en-3-one, 1s,4r-3-azabicyclo 2.2.1 hept-5-en-2-one, 2-azabicyclo 2,2,1 hept-5-en-3-one, vince lactam, 2-azabicyclo 2.2.1 hept-5-en-3-one, 1r,4s, --2-azabicyclo 2.2.1 hept-5-en-3-one, --vince lactam, 1s-+-2-azabicyclo 2.2.1 hept-5-en-3-one
PubChem CID 2725037
IUPAC Name (1S,4R)-3-azabicyclo[2.2.1]hept-5-en-2-one
SMILES C1C2C=CC1NC2=O

Specifications

Absorbance (c=1, CH2Cl2) (1 cm cell), (c=1, CH2Cl2) (1 cm cell) at 0nm, 0.1 max. at 400nm
CAS Min % 97.5
CAS Max % 100.0
Melting Point 92°C to 100°C
Color White to Beige
Enantiomeric Excess 95% min. (HPLC)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation -560
Formula Weight 109.13
Percent Purity 98%,>95% ee
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (-)-2-Azabicyclo[2.2.1]hept-5-en-3-one, >95%ee
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (-)-2-Azabicyclo¢2.2.1!hept-5-en-3-one
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.