missing translation for 'onlineSavingsMsg'
Learn More

2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%, Alfa Aesar™

Product Code. 15432168
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15432168 1 g 1g
15442168 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15432168 Supplier Thermo Scientific Alfa Aesar Supplier No. H55812.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 49805-30-3
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00213364
InChI Key DDUFYKNOXPZZIW-UHFFFAOYSA-N
Synonym 2-azabicyclo 2.2.1 hept-5-en-3-one, +/--2-azabicyclo 2.2.1 hept-5-en-3-one, 3-azabicyclo 2.2.1 hept-5-en-2-one, inverted exclamation marka-2-azabicyclo 2.2.1 hept-5-en-3-one, vince lactam, 1s-+-2-azabicyclo 2.2.1 hept-5-en-3-one, --vince lactam, --2-azabicyclo 2.2.1 hept-5-en-3-one, +-2-azabicyclo 2.2.1 hept-5-en-3-one
PubChem CID 548678
IUPAC Name 3-azabicyclo[2.2.1]hept-5-en-2-one
SMILES C1C2C=CC1NC2=O

Specifications

Melting Point 54°C to 58°C
Boiling Point 102°C to 106°C (0.25 mmHg)
Flash Point 110°C (230°F)
Quantity 1 g
Beilstein 508342
Formula Weight 109.13
Percent Purity 98%
Chemical Name or Material 2-Azabicyclo[2.2.1]hept-5-en-3-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.