missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ 2-APB

Product Code. 18075342
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18075342 10 mg 10mg
18034702 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18075342 Supplier R&D Systems™ Supplier No. 1224/10
  • ONLINE EXCLUSIVE PRICE
    £96.00
    £90.30 / 10mg
    Save £5.70
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. IP3 receptor antagonist. Also TRP channel modulator

A functional and membrane permeable D-myo-inositol 1,4,5-trisphosphate (IP3) receptor antagonist (IC50 = 42 μM). Stimulates store-operated calcium (SOC) release at low concentrations (< 10 μM) and inhibits it at higher concentrations (up to 50 μM). Increases STIM-Orai channel conductance and limits ion selectivity. Modulator of TRP channels; blocks TRPC1, TRPC3, TRPC5, TRPC6, TRPV6, TRPM3, TRPM7, TRPM8 and TRPP2 and at higher concentrations stimulates TRPV1, TRPV2 and TRPV3. Also blocks specific gap channel subtypes.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 524-95-8
Molecular Formula C14H16BNO

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 225.1
Purity >98%
Molecular Weight (g/mol) 225.10
Chemical Name or Material 2-APB
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.