missing translation for 'onlineSavingsMsg'
Learn More

2-Anthracenecarboxylic Acid, TRC

Product Code. 30329896
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30329896 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329896 Supplier Toronto Research Chemicals Supplier No. TRCA69781350MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Anthracenecarboxylic Acid
CAS 613-08-1
Molecular Formula C15H10O2
Quantity 50 mg
InChI Formula InChI=1S/C15H10O2/c16-15(17)13-6-5-12-7-10-3-1-2-4-11(10)8-14(12)9-13/h1-9H,(H,16,17)
SMILES OC(=O)c1ccc2cc3ccccc3cc2c1
IUPAC Name anthracene-2-carboxylic acid
Molecular Weight (g/mol) 222.23
Formula Weight 222.0681
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.