missing translation for 'onlineSavingsMsg'
Learn More

2-Anilinopyrimidine-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10307314
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10307314 1 g Amber Glass Bottle 1g
10709425 250 mg Amber Glass Bottle 250mg
10158754 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10307314 Supplier Thermo Scientific Maybridge Supplier No. CC69204DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 108002-87-5
Molecular Formula C11H9N3O
Molecular Weight (g/mol) 199.213
InChI Key QNFHXIZSGIGYOU-UHFFFAOYSA-N
Synonym 2-phenylamino pyrimidine-5-carbaldehyde, 2-anilinopyrimidine-5-carboxaldehyde, 2-phenylamino pyrimidine-5-carboxaldehyde, 2-anilinopyrimidine-5-carbaldehyde
PubChem CID 33589542
IUPAC Name 2-anilinopyrimidine-5-carbaldehyde
SMILES C1=CC=C(C=C1)NC2=NC=C(C=N2)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 199.21
Percent Purity 97%
Chemical Name or Material 2-anilinopyrimidine-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.