missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminothiophenol 90%, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 2-Aminothiophenol |
| CAS | 137-07-5 |
| Molecular Formula | C6H7NS |
| Quantity | 5 g |
| Synonym | o-Aminobenzenethiol, 1-Amino-2-mercaptobenzene, 2-Amino-1-mercaptobenzene, 2-Aminobenzenethiol, 2-Aminophenyl Mercaptan, 2-Aminothiophenol, 2-Mercaptoaniline, 2-Thioaniline, NSC 106635, o-Aminobenzenethiol, o-Aminophenyl Mercaptan, o-Aminothiophenol, o-Mercaptoaniline, alpha-Aminothiophenol |
| InChI Formula | InChI=1S/C6H7NS/c7-5-3-1-2-4-6(5)8/h1-4,8H,7H2 |
| SMILES | Nc1ccccc1S |
| IUPAC Name | 2-aminobenzenethiol |
| Molecular Weight (g/mol) | 125.19 |
| Formula Weight | 125.0299 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction