missing translation for 'onlineSavingsMsg'
Learn More

2-Aminothiobenzamide, 97%

Product Code. p-7034149
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15478267 1 g 1g
15488267 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 15478267

Brand: Thermo Scientific Alfa Aesar H51809.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2454-39-9 | C7H8N2S | 152.215 g/mol

It can be heated to produce 2-trifluoromethyl-3H-quinazoline-4-thione in the presence of reagent molecular sieves and solvent CHCl3 with reaction time of 2 hours. It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be heated to produce 2-trifluoromethyl-3H-quinazoline-4-thione in the presence of reagent molecular sieves and solvent CHCl3 with reaction time of 2 hours. It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2454-39-9
Molecular Formula C7H8N2S
Molecular Weight (g/mol) 152.215
MDL Number MFCD00963496
InChI Key HPZKAJRFABCGFF-UHFFFAOYSA-N
Synonym 2-aminothiobenzamide, 2-amino-thiobenzamide, 2-aminobenzothioamide, benzenecarbothioamide, 2-amino, thioanthranilamide, o-aminothiobenzamide, 2-aminobenzene-1-carbothioamide, amino 2-aminophenyl methane-1-thione, benzamide, o-aminothio, thioanthranilsaureamid
PubChem CID 1550990
IUPAC Name 2-aminobenzenecarbothioamide
SMILES C1=CC=C(C(=C1)C(=S)N)N

Specifications

Melting Point 115°C to 120°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 152.21
Percent Purity 97%
Chemical Name or Material 2-Aminothiobenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.