missing translation for 'onlineSavingsMsg'
Learn More

2-Aminothiazole-5-carboxylic acid, 95%

Product Code. 10301280 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10301280 5 g Glass bottle 5g
10402590 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10301280 Supplier Thermo Scientific Acros Supplier No. 429570050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40283-46-3 | C4H4N2O2S | 144.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40283-46-3
Molecular Formula C4H4N2O2S
Molecular Weight (g/mol) 144.15
MDL Number MFCD06203554
InChI Key ZFMRDDYYJJCBKC-UHFFFAOYSA-N
Synonym 2-aminothiazole-5-carboxylic acid, 2-amino-5-thiazolecarboxylic acid, 5-thiazolecarboxylic acid, 2-amino, 2-amino-5-carboxy-1,3-thiazole, pubchem21384, acmc-209jcp, 2-aminothiazole-5-carboxylicacid, 5-thiazolecarboxylicacid, 2-amino, 2-aminothiazole-5-formic acid, 2-azanyl-1,3-thiazole-5-carboxylic acid
PubChem CID 315243
IUPAC Name 2-amino-1,3-thiazole-5-carboxylic acid
SMILES C1=C(SC(=N1)N)C(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 182.0°C to 184.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 144.15
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminothiazole-5-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Aminothiazole-5-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.