missing translation for 'onlineSavingsMsg'
Learn More

2-Aminothiazole-4-acetic Acid, Mikromol™

Product Code. 30051637
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051637 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051637 Supplier Mikromol Supplier No. MM3776.010025
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-Aminothiazole-4-acetic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Aminothiazole-4-acetic Acid
CAS 29676-71-9
Analyte or Component Names 2-Aminothiazole-4-acetic Acid
Shipping Condition Ice pack (-20°C)
Molecular Formula C5H6N2O2S
Certifications/Compliance ISO 9001
Synonym (2-Amino-1,3-thiazol-4-yl)acetic acid, (2-Amino-4-thiazoyl)acetic acid, 2-(2-Amino-4-thiazole)acetic acid, 2-(2-Amino-4-thiazolyl)acetic acid, 2-Amino-4-thiazolylacetic acid, 2-Aminothiazol-4-acetic acid, 2-Aminothiazole-4-acetic acid, SQ 272, Ceftibuten Related Impurity 2, Mirabegron Impurity 7
InChI Formula InChI=1S/C5H6N2O2S/c6-5-7-3(2-10-5)1-4(8)9/h2H,1H2,(H2,6,7)(H,8,9)
SMILES Nc1nc(CC(=O)O)cs1
IUPAC Name 2-(2-aminothiazol-4-yl)acetic acid
Molecular Weight (g/mol) 158.18
Quantity 25 mg
Formula Weight 158.015 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.