missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopyrimidine, 98%

Product Code. 11420448
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420448 100 g 100g
11430448 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420448 Supplier Thermo Scientific Alfa Aesar Supplier No. B24594.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-12-6 | C4H5N3 | 95.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-12-6
Molecular Formula C4H5N3
Molecular Weight (g/mol) 95.11
MDL Number MFCD00006089
InChI Key LJXQPZWIHJMPQQ-UHFFFAOYSA-N
Synonym 2-aminopyrimidine, 2-pyrimidinamine, pyrimidin-2-ylamine, pyrimidinamine, aminopyrimidine, 2-pyrimidiylamine, pyrimidine, 2-amino, 2-pyridiylamine, 1,2-dihydro-2-iminopyrimidine, pyrimidine, amino
PubChem CID 7978
ChEBI CHEBI:38618
IUPAC Name pyrimidin-2-amine
SMILES NC1=NC=CC=N1

Specifications

Melting Point 123°C to 127°C
Boiling Point 159°C (186 mmHg)
Quantity 100 g
Beilstein 107014
Formula Weight 95.11
Percent Purity 98%
Chemical Name or Material 2-Aminopyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.