missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopyridine, TRC

Product Code. 30005157
Change view
Click to view available options
Quantity:
1 g
50 g
250 g
Unit Size:
1g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005157 50 g 50g
30004929 250 g 250g
18002000 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005157 Supplier Toronto Research Chemicals Supplier No. TRCA62898550G
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-29-0
Molecular Formula C5 H6 N2
Molecular Weight (g/mol) 94.11
Synonym Pyridine, 2-amino- (8CI), 1,2-Dihydro-2-iminopyridine, 2-Aminopyridine, 2-Pyridinylamine, 2-Pyridylamine, NSC 431, o-Aminopyridine, α-Aminopyridine, α-Pyridinamine, α-Pyridylamine
IUPAC Name pyridin-2-amine
SMILES Nc1ccccn1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C5H6N2/c6-5-3-1-2-4-7-5/h1-4H,(H2,6,7)
Formula Weight 94.0531
Percent Purity >95%
Chemical Name or Material 2-Aminopyridine
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.