missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopyridine, 99+%

Product Code. 10695512 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10695512 500 g Plastic bottle 500g
10519440 100 g Plastic bottle 100g
10445555 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10695512 Supplier Thermo Scientific Acros Supplier No. 104545000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 504-29-0 | C5H6N2 | 94.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-29-0
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.12
MDL Number MFCD00006312
InChI Key ICSNLGPSRYBMBD-UHFFFAOYSA-N
Synonym 2-aminopyridine, 2-pyridinamine, 2-pyridylamine, aminopyridine, pyridinamine, o-aminopyridine, 2-aminopyridin, 2-aminopryidine, amino-2 pyridine, pyridine, 2-amino
PubChem CID 10439
IUPAC Name pyridin-2-amine
SMILES C1=CC=NC(=C1)N

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 55°C to 60°C
Color Beige-Yellow to Cream
Boiling Point 204°C to 210°C
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula (C5H4N)NH2
Quantity 500 g
Beilstein 22,428
Fieser 09,18
Merck Index 15,469
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,benzene,ether and hot,petroleum ether,sparingly soluble in toluene
Formula Weight 94.12
Percent Purity 99+%
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material 2-Aminopyridine, p.a.
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.