missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopyridine, 99%

Product Code. 11409523
Change view
Click to view available options
Quantity:
25 g
250 g
1000 g
Unit Size:
1000g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409523 250 g 250g
11359679 25 g 25g
11499513 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409523 Supplier Thermo Scientific Alfa Aesar Supplier No. A12374.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 504-29-0 | C5H6N2 | 94.117 g/mol

A compound has been shown to reversibly block voltage-dependent potassium channels. Also a common impurity from the synthesis of compounds found in hair dye. Used as a water miscible solvent and as an intermediate in manufacturing pharmaceuticals like diuretics. It is used as a research tool, in characterizing subtypes of the potassium channel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A compound has been shown to reversibly block voltage-dependent potassium channels. Also a common impurity from the synthesis of compounds found in hair dye. Used as a water miscible solvent and as an intermediate in manufacturing pharmaceuticals like diuretics. It is used as a research tool, in characterizing subtypes of the potassium channel.

Solubility
Slightly soluble in water(8g/100 mL at 19°C)

Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-29-0
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.117
MDL Number MFCD00006312
InChI Key ICSNLGPSRYBMBD-UHFFFAOYSA-N
Synonym 2-aminopyridine, 2-pyridinamine, 2-pyridylamine, aminopyridine, pyridinamine, o-aminopyridine, 2-aminopyridin, 2-aminopryidine, amino-2 pyridine, pyridine, 2-amino
PubChem CID 10439
IUPAC Name pyridin-2-amine
SMILES C1=CC=NC(=C1)N

Specifikationer

Melting Point 56°C to 60°C
Boiling Point 209°C to 211°C
Flash Point 92°C (197°F)
Quantity 250 g
UN Number UN2671
Beilstein 105785
Sensitivity Hygroscopic
Merck Index 14,473
Solubility Information Slightly soluble in water(8g/100 mL at 19°C)
Formula Weight 94.12
Percent Purity 99%
Chemical Name or Material 2-Aminopyridine
Visa mer Visa mindre
compliance-icons compliance-icons
Produktbeteckning
  • 2-Aminopyridine
Signalord
  • Danger
Riskkategorier
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Faroangivelser
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Skyddsangivelse
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Kompletterande information
  • MIXTURE LIST-Contain: 2-Aminopyridine

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.