missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminopyridine-6-methanol, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
4-Aminopyridine (4-AP), a well-known potassium channel blocker, has been shown to enhance conduction in injured and demyelinated axons. Aminopyridine has a narrow therapeutic range in clinical application. In the current study, we investigated whether AP-MeOH can enhance impulse conduction in an animal model of MS.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Slightly soluble.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Slightly soluble.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | 87°C to 92°C |
Quantity | 1 g |
Solubility Information | Slightly soluble. |
Formula Weight | 124.14 |
Percent Purity | 97% |
Chemical Name or Material | 2-Aminopyridine-6-methanol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'