missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopyridine-4-carboxylic acid, 96%, Thermo Scientific™

Product Code. 11379835
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379835 25 g 25g
11359835 1 g 1g
11369835 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11379835 Supplier Thermo Scientific Alfa Aesar Supplier No. L19845.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13362-28-2
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD00233714
InChI Key QMKZZQPPJRWDED-UHFFFAOYSA-N
Synonym 2-aminoisonicotinic acid, 2-amino-4-pyridinecarboxylic acid, 2-amino-4-pyridine carboxylic acid, 2-amino-isonicotinic acid, 4-pyridinecarboxylic acid, 2-amino, 2-amino isonicotinic acid, pubchem2426, 2-aminoisonicotinicacid, acmc-209bt5, ksc174m9j
PubChem CID 278396
IUPAC Name 2-aminopyridine-4-carboxylic acid
SMILES C1=CN=C(C=C1C(=O)O)N

Specifications

Melting Point 300°C to 302°C
Odor Odorless
Quantity 25 g
Beilstein 3156
Formula Weight 138.13
Percent Purity 96%
Chemical Name or Material 2-Aminopyridine-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.