missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopurine, TRC

Product Code. 30312303
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30312303 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312303 Supplier Toronto Research Chemicals Supplier No. TRCA628918250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Aminopurine
CAS 452-06-2
Purity Grade Notes HPLC
Molecular Formula C5 H5 N5
Quantity 250 mg
Synonym 9H-Purin-2-amine, 1H-Purin-2-amine (9CI), Purine, 2-amino- (6CI,8CI), (9H-Purin-2-yl)amine, 2-Aminopurine, Isoadenine, NSC 24129, SQ 22451, 2-Amino-9H-purine
InChI Formula InChI=1S/C5H5N5/c6-5-7-1-3-4(10-5)9-2-8-3/h1-2H,(H3,6,7,8,9,10)
SMILES Nc1ncc2nc[nH]c2n1
IUPAC Name 9H-purin-2-amine
Molecular Weight (g/mol) 135.13
Formula Weight 135.0545
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.