missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopurine, Mikromol™

Product Code. 30050298
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050298 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050298 Supplier Mikromol Supplier No. MM0620.190025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-Aminopurine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Aminopurine
CAS 452-06-2
Analyte or Component Names 2-Aminopurine
Shipping Condition Room Temperature
Molecular Formula C5 H5 N5
Certifications/Compliance ISO 9001
Synonym 9H-Purin-2-amine, 1H-Purin-2-amine (9CI), Purine, 2-amino- (6CI,8CI), (9H-Purin-2-yl)amine, Isoadenine, NSC 24129, SQ 22451, 2-Amino-9H-purine, Famciclovir Related Compound E
InChI Formula InChI=1S/C5H5N5/c6-5-7-1-3-4(10-5)9-2-8-3/h1-2H,(H3,6,7,8,9,10)
SMILES Nc1ncc2nc[nH]c2n1
IUPAC Name 9H-purin-2-amine
Molecular Weight (g/mol) 135.13
Quantity 25 mg
Formula Weight 135.0545 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.