missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopurine, 98%

Product Code. 15416649
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15416649 500 mg 500mg
15406649 250 mg 250mg
15496639 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15416649 Supplier Thermo Scientific Alfa Aesar Supplier No. J64919.ME
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A protein kinase R inhibitor | CAS: 452-06-2 | C5H5N5 | 135.13 g/mol

A protein kinase R inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-06-2
Molecular Formula C5H5N5
Molecular Weight (g/mol) 135.13
MDL Number MFCD00005566
InChI Key MWBWWFOAEOYUST-UHFFFAOYSA-N
Synonym 2-aminopurine, 1h-purin-2-amine, 9h-purin-2-amine, purine, 2-amino, aminopurine, 2-amino purine, 2'-amino-purine, unii-o14b3u97fw, ccris 759, 9h-purin-2-ylamine
PubChem CID 9955
ChEBI CHEBI:479072
IUPAC Name 7H-purin-2-amine
SMILES NC1=NC=C2NC=NC2=N1

Specifications

Melting Point 280°C to 282°C
Color White to Yellow
Odor Odorless
Quantity 500 mg
Formula Weight 135.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Aminopurine
What is the solubility of 2-Aminopurine 98%?

We have the following information regarding solubility for 2-Aminopurin 98%:
- Soluble in a formic acid and water solution (1:1 v/v) at 50 mg/ml by heating.
- Soluble in a glacial acetic acid and PBS solution (200:1) by heating at 60 degrees C for an 150 mM solution. This solution can then be aliquoted and stored frozen. The aliquots should be thawed, heated and mixed before usage.

What is the molecular weight of 2-Aminopurine 98%?

The molecular weight is 135.13 g/mol (this product is the non-salt form, CAS 452-06-2).

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.