Learn More
2-Aminopurine, 98%
Description
A protein kinase R inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 280°C to 282°C |
| Color | White to Yellow |
| Odor | Odorless |
| Quantity | 250 mg |
| Formula Weight | 135.13 |
| Percent Purity | 98% |
| Physical Form | Powder |
| Chemical Name or Material | 2-Aminopurine |
Frequently Asked Questions (FAQs)
We have the following information regarding solubility for 2-Aminopurin 98%:
- Soluble in a formic acid and water solution (1:1 v/v) at 50 mg/ml by heating.
- Soluble in a glacial acetic acid and PBS solution (200:1) by heating at 60 degrees C for an 150 mM solution. This solution can then be aliquoted and stored frozen. The aliquots should be thawed, heated and mixed before usage.
The molecular weight is 135.13 g/mol (this product is the non-salt form, CAS 452-06-2).
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.