missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminophenylboronic acid hydrochloride, 97%, Thermo Scientific Chemicals
£79.70 - £287.00
Chemical Identifiers
CAS | 863753-30-4 |
---|---|
Molecular Formula | C6H9BClNO2 |
Molecular Weight (g/mol) | 173.40 |
MDL Number | MFCD02258096 |
InChI Key | WPDASZCYRKGSTO-UHFFFAOYSA-N |
Synonym | 2-aminophenylboronic acid hydrochloride, 2-aminophenyl boronic acid hydrochloride, 2-aminophenylboronic acid hcl, 2-aminophenylboronic acid, hydrochloride, 2-2-aminophenyl boronic acid hydrochloride, boronic acid, 2-aminophenyl-, hydrochloride, pubchem1746, ksc914i9f, 2-aminophenylboronic acid,hcl, 2-aminophenylboronic acid, hcl |
PubChem CID | 16427090 |
SMILES | Cl.NC1=CC=CC=C1B(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11996121
|
Thermo Scientific Acros
445310010 |
1 g |
£79.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11906131
|
Thermo Scientific Acros
445310050 |
5 g |
£287.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
863753-30-4 | |
173.40 | |
WPDASZCYRKGSTO-UHFFFAOYSA-N | |
16427090 |
C6H9BClNO2 | |
MFCD02258096 | |
2-aminophenylboronic acid hydrochloride, 2-aminophenyl boronic acid hydrochloride, 2-aminophenylboronic acid hcl, 2-aminophenylboronic acid, hydrochloride, 2-2-aminophenyl boronic acid hydrochloride, boronic acid, 2-aminophenyl-, hydrochloride, pubchem1746, ksc914i9f, 2-aminophenylboronic acid,hcl, 2-aminophenylboronic acid, hcl | |
Cl.NC1=CC=CC=C1B(O)O |
Specifications
863753-30-4 | |
Glass Bottle | |
MFCD02258096 | |
WPDASZCYRKGSTO-UHFFFAOYSA-N | |
(2-aminophenyl)boronic acid hydrochloride | |
16427090 | |
97% |
96% min. (HPLC) | |
C6H9BClNO2 | |
2-aminophenylboronic acid hydrochloride, 2-aminophenyl boronic acid hydrochloride, 2-aminophenylboronic acid hcl, 2-aminophenylboronic acid, hydrochloride, 2-2-aminophenyl boronic acid hydrochloride, boronic acid, 2-aminophenyl-, hydrochloride, pubchem1746, ksc914i9f, 2-aminophenylboronic acid,hcl, 2-aminophenylboronic acid, hcl | |
Cl.NC1=CC=CC=C1B(O)O | |
173.40 | |
173.41 | |
2-Aminophenylboronic acid hydrochloride |
Safety and Handling
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2-Aminophenylboronic acid hydrochloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only