missing translation for 'onlineSavingsMsg'
Learn More

2-Aminophenyl Beta-D-Glucuronide, TRC

Product Code. 30012898
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012898 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30012898 Supplier Toronto Research Chemicals Supplier No. TRCA62399550MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 15959-03-2
Molecular Formula C12H15NO7
Quantity 50 mg
Synonym o-Aminophenyl beta-D-Glucopyranosiduronic Acid, 2-Aminophenol Glucopyranuronoside, 2-Aminophenol Glucuronide, NSC 89270;
InChI Formula InChI=1S/C12H15NO7/c13-5-3-1-2-4-6(5)19-12-9(16)7(14)8(15)10(20-12)11(17)18/h1-4,7-10,12,14-16H,13H2,(H,17,18)/t7-,8-,9+,10-,12+/m0/s1
SMILES Nc1ccccc1O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O
IUPAC Name (2S,3S,4S,5R,6S)-6-(2-aminophenoxy)-3,4,5-trihydroxyoxane-2-carboxylic acid
Molecular Weight (g/mol) 285.25
Formula Weight 285.0849
Chemical Name or Material 2-Aminophenyl beta-D-Glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.