missing translation for 'onlineSavingsMsg'
Learn More

2-Aminophenol, TRC

Product Code. 30325037
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30325037 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30325037

Brand: Toronto Research Chemicals TRCA61893010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Aminophenol
CAS 95-55-6
Molecular Formula C6 H7 N O
Quantity 10 g
Synonym Phenol, o-amino- (8CI), 2-Aminophenol, (2-Hydroxyphenyl)amine, 1-Amino-2-hydroxybenzene, 1-Hydroxy-2-aminobenzene, 2-Amino-1-hydroxybenzene, 2-Aminophenyl alcohol, 2-Hydroxyaniline, 2-Hydroxybenzenamine, BASF Ursol 3GA, Benzofur GG, C.I. 76520, C.I. Oxidation Base 17, Fouramine OP, NSC 1534, NSC 226261, Nako Yellow 3GA, Paradone Olive Green B, Pelagol 3GA, Pelagol Grey GG, Rodol 2G, Zoba 3GA, o-Aminohydroxybenzene, o-Aminophenol, o-Hydroxyaniline, o-Hydroxyphenylamine
InChI Formula InChI=1S/C6H7NO/c7-5-3-1-2-4-6(5)8/h1-4,8H,7H2
SMILES Nc1ccccc1O
IUPAC Name 2-aminophenol
Molecular Weight (g/mol) 109.13
Formula Weight 109.0528
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.