missing translation for 'onlineSavingsMsg'
Learn More

2-Aminonicotinic acid, 98%

Product Code. 10409780 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10409780 25 g Glass bottle 25g
10000680 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10409780 Supplier Thermo Scientific Acros Supplier No. 104160250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5345-47-1 | C6H6N2O2 | 138.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5345-47-1
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00006318
InChI Key KPIVDNYJNOPGBE-UHFFFAOYSA-N
Synonym 2-aminonicotinic acid, 2-amino-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-amino, 2-amino-nicotinic acid, 2-amino-3-carboxypyridine, nicotinic acid, 2-amino, aminonicotinic acid, h-nh 2 nicot-oh, amino-nicotinic acid, 2-aminonicotinicacid
PubChem CID 72930
ChEBI CHEBI:68572
IUPAC Name 2-aminopyridine-3-carboxylic acid
SMILES NC1=NC=CC=C1C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 295.0°C to 297.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula H2N(C5H3N)COOH
Quantity 25 g
Beilstein 22, 542
Solubility Information Solubility in water: soluble.
Formula Weight 138.13
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminonicotinic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Aminonicotinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.