missing translation for 'onlineSavingsMsg'
Learn More
Learn More

2-Aminonicotinic acid, 98%, Thermo Scientific™
Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Color | White to Yellow |
Melting Point | 295.0°C to 297.0°C |
Assay Percent Range | 97.5% min. (HPLC) |
Infrared Spectrum | Authentic |
Linear Formula | H2N(C5H3N)COOH |
Beilstein | 22, 542 |
Packaging | Glass bottle |
Quantity | 5g |
Show More |
Safety and Handling

Product Identifier
- 2-Aminonicotinic acid
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only