missing translation for 'onlineSavingsMsg'
Learn More

2-(Aminomethyl)pyrazine, TRC

Product Code. 30320968
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30320968 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320968 Supplier Toronto Research Chemicals Supplier No. TRCA6174405G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Aminomethyl)pyrazine
CAS 20010-99-5
Molecular Formula C5 H7 N3
Quantity 5 g
Synonym (Aminomethyl)-pyrazine, Pyrazinemethanamine, NSC 105567, Pyrazin-2-ylmethanamine, [(Pyrazin-2-yl)methyl]amine
InChI Formula InChI=1S/C5H7N3/c6-3-5-4-7-1-2-8-5/h1-2,4H,3,6H2
SMILES NCc1cnccn1
IUPAC Name pyrazin-2-ylmethanamine
Molecular Weight (g/mol) 109.129
Formula Weight 109.064
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.