missing translation for 'onlineSavingsMsg'
Learn More

2-(Aminomethyl)pyrazine, 95%

Product Code. 15254328 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15254328 250 mg 250mg
15244328 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15254328 Supplier Thermo Scientific Acros Supplier No. 454162500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20010-99-5 | C5H7N3 | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20010-99-5
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.13
MDL Number MFCD00673149
InChI Key HQIBSDCOMQYSPF-UHFFFAOYSA-N
Synonym 2-aminomethylpyrazine, 2-pyrazinemethanamine, 2-aminomethyl pyrazine, pyrazin-2-yl methanamine, 1-pyrazin-2-ylmethanamine, pyrazinemethanamine, 2-aminomethyl-pyrazine, pyrazin-2-ylmethylamine, pyrazin-2-ylmethyl amine
PubChem CID 266781
IUPAC Name pyrazin-2-ylmethanamine
SMILES NCC1=CN=CC=N1

Specifications

Boiling Point 87.0°C to 88.0°C (3.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Quantity 250 mg
Formula Weight 109.13
Percent Purity 95%
Chemical Name or Material 2-(Aminomethyl)pyrazine
compliance-icons compliance-icons
Product Identifier
  • 2-(Aminomethyl)pyrazine, 95%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.