missing translation for 'onlineSavingsMsg'
Learn More

2-Aminomethyl-1H-imidazole dihydrochloride, 97%

Product Code. 15274368 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15274368 1 g 1g
15284368 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15274368 Supplier Thermo Scientific Acros Supplier No. 454590010
  • ONLINE EXCLUSIVE PRICE
    £64.20
    £57.65 / 1g
    Save £6.55
    10% Off
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 22600-77-7 | C4H9Cl2N3 | 170.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22600-77-7
Molecular Formula C4H9Cl2N3
Molecular Weight (g/mol) 170.04
MDL Number MFCD06738779
InChI Key KYUDBQDDNKPSIC-UHFFFAOYSA-N
Synonym 1h-imidazol-2-yl methanamine dihydrochloride, 2-aminomethylimidazole dihydrochloride, 2-aminomethyl-1h-imidazole dihydrochloride, 1h-imidazol-2-ylmethylamine dihydrochloride, 1h-imidazol-2-ylmethanamine dihydrochloride, 1h-imidazole-2-methanamine dihydrochloride, 1h-imidazole-2-methanamine, dihydrochloride, 1-1h-imidazol-2-yl methanamine dihydrochloride, 1h-imidazol-2-ylmethyl amine dihydrochloride, 2-aminomethylimidazole hcl
PubChem CID 12417863
SMILES Cl.Cl.NCC1=NC=CN1

Specifications

Melting Point 250.0°C to 251.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 to max. 102.5% min. (Argentometry)
Packaging Glass Bottle
Quantity 1 g
IUPAC Name 1-(1H-imidazol-2-yl)methanamine dihydrochloride
Formula Weight 170.04
Percent Purity 98%
Chemical Name or Material 2-Aminomethyl-1H-imidazole dihydrochloride
compliance-icons
Product Identifier
  • 2-Aminomethyl-1H-imidazole dihydrochloride, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.