missing translation for 'onlineSavingsMsg'
Learn More

2-(Aminomethyl)-1-ethylpyrrolidine, 95%

Product Code. 10032330 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10032330 10 g Glass bottle 10g
10761261 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10032330 Supplier Thermo Scientific Acros Supplier No. 179480100
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 26116-12-1 | C7H16N2 | 128.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26116-12-1
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.22
MDL Number MFCD00003178
InChI Key UNRBEYYLYRXYCG-UHFFFAOYSA-N
Synonym 2-aminomethyl-1-ethylpyrrolidine, 1-ethylpyrrolidin-2-yl methanamine, n-ethyl-2-aminomethylpyrrolidine, 2-pyrrolidinemethanamine, 1-ethyl, 1-ethylpyrrolidin-2-ylmethylamine, 1-ethylpyrrolidin-2-yl methylamine, 1-ethyl-2-aminomethylpyrrolidine, 1-ethyl-2-aminomethyl pyrrolidine, 1-1-ethylpyrrolidin-2-yl methanamine
PubChem CID 117295
IUPAC Name (1-ethylpyrrolidin-2-yl)methanamine
SMILES CCN1CCCC1CN

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Density 0.8840g/mL
Boiling Point 58.0°C to 60.0°C (16.0 mmHg)
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Refractive Index 1.4655 to 1.4675
Quantity 10 g
Specific Gravity 0.884
Formula Weight 128.22
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-(Aminomethyl)-1-ethylpyrrolidine, 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-(Aminomethyl)-1-ethylpyrrolidine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.