missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminomethyl-1-Boc-piperidine, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Reactant for synthesis of: TRPV1 antagonists, CHK1 inhibitors, p38a MAP kinase inhibitors, Carbamoylphosphonates, Macrolactams via carbonylation/intramolecular amidation, Intramolecular transamidation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reactant for synthesis of: TRPV1 antagonists, CHK1 inhibitors, p38a MAP kinase inhibitors, Carbamoylphosphonates, Macrolactams via carbonylation/intramolecular amidation, Intramolecular transamidation.
Solubility
Soluble in DMSO.
Notes
Refrigerated (2-8°C). Incompatible with oxidizing agents.
Reactant for synthesis of: TRPV1 antagonists, CHK1 inhibitors, p38a MAP kinase inhibitors, Carbamoylphosphonates, Macrolactams via carbonylation/intramolecular amidation, Intramolecular transamidation.
Solubility
Soluble in DMSO.
Notes
Refrigerated (2-8°C). Incompatible with oxidizing agents.
Specifications
Density | 1.012 |
Flash Point | >110°C (230°F) |
Refractive Index | 1.4745 |
Quantity | 250 mg |
UN Number | UN3082 |
Solubility Information | Soluble in DMSO. |
Formula Weight | 214.31 |
Percent Purity | 95% |
Chemical Name or Material | 2-Aminomethyl-1-Boc-piperidine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'