missing translation for 'onlineSavingsMsg'
Learn More

2-Aminomethyl-1-Boc-piperidine, 95%

Product Code. 15420798
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15420798 5 g 5g
15430798 250 mg 250mg
15410798 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15420798 Supplier Thermo Scientific Alfa Aesar Supplier No. H54507.06
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 370069-31-1 | C11H22N2O2 | 214.309 g/mol

Reactant for synthesis of: TRPV1 antagonists, CHK1 inhibitors, p38a MAP kinase inhibitors, Carbamoylphosphonates, Macrolactams via carbonylation/intramolecular amidation, Intramolecular transamidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant for synthesis of: TRPV1 antagonists, CHK1 inhibitors, p38a MAP kinase inhibitors, Carbamoylphosphonates, Macrolactams via carbonylation/intramolecular amidation, Intramolecular transamidation.

Solubility
Soluble in DMSO.

Notes
Refrigerated (2-8°C). Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 370069-31-1
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.309
MDL Number MFCD01317791
InChI Key PTVRCUVHYMGECC-UHFFFAOYSA-N
Synonym 2-aminomethyl-1-boc-piperidine, tert-butyl 2-aminomethyl piperidine-1-carboxylate, 2-aminomethyl-1-n-boc-piperidine, 1-boc-2-aminomethyl-piperidine, 1-boc-2-aminomethyl piperidine, 1-boc-2-aminomethylpiperidine, 2-aminomethyl-piperidine-1-carboxylic acid tert-butyl ester, 1-n-boc-2-aminomethyl piperidine
PubChem CID 2756482
IUPAC Name tert-butyl 2-(aminomethyl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCCC1CN

Specifications

Density 1.012
Flash Point >110°C (230°F)
Refractive Index 1.4745
Quantity 5 g
UN Number UN3082
Solubility Information Soluble in DMSO.
Formula Weight 214.31
Percent Purity 95%
Chemical Name or Material 2-Aminomethyl-1-Boc-piperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.