missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoisobutyric acid methyl ester hydrochloride, 99%

Product Code. 11405980
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11405980

Brand: Thermo Scientific Alfa Aesar H26550.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15028-41-8 | C5H12ClNO2 | 153.606 g/mol

Methyl α-aminoisobutyrate is used as a specific substrate of amino acid transport system and in studies on amino acid transport systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl α-aminoisobutyrate is used as a specific substrate of amino acid transport system and in studies on amino acid transport systems.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15028-41-8
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.606
MDL Number MFCD00214247
InChI Key NVWZNEDLYYLQJC-UHFFFAOYSA-N
Synonym methyl 2-amino-2-methylpropanoate hydrochloride, methyl 2-aminoisobutyrate hydrochloride, h-aib-ome.hcl, alpha-aminoisobutyric acid methyl ester hydrochloride, 2-aminoisobutyric acid methyl ester hydrochloride, methyl alpha-aminoisobutyrate hydrochloride, methyl 2-methylalaninate hydrochloride, alanine, 2-methyl-, methyl ester, hydrochloride, aib-ome hydrochloride, pubchem16472
PubChem CID 13258034
IUPAC Name methyl 2-amino-2-methylpropanoate;hydrochloride
SMILES CC(C)(C(=O)OC)N.Cl

Specifications

Melting Point ∽185°C (decomposition)
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 153.61
Percent Purity 99%
Chemical Name or Material 2-Aminoisobutyric acid methyl ester hydrochloride
compliance-icons
Product Identifier
  • 2-Aminoisobutyric acid methyl ester hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Methyl 2-methylalaninate-hydrogen chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.