missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (+/-)-2-Aminoheptane sulfate, 99%, Thermo Scientific™

Product Code. 11352596
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352596 25 g 25g
11362596 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352596 Supplier Alfa Aesar™ Supplier No. L08596.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6411-75-2
Molecular Formula C14H36N2O4S
Molecular Weight (g/mol) 328.512
MDL Number MFCD00050844
InChI Key XKUUMWKWUZRRPD-UHFFFAOYSA-N
Synonym sulfuric acid; bis tuaminoheptane, dsstox_cid_25845, dsstox_rid_81167, dsstox_gsid_45845, -2-aminoheptane sulfate, tuaminoheptane sulfate usp, heptan-2-amine sulfate 2:1, ^+-2-aminoheptane sulfate
PubChem CID 44134533
IUPAC Name heptan-2-amine;sulfuric acid
SMILES CCCCCC(C)N.CCCCCC(C)N.OS(=O)(=O)O

Specifications

Melting Point 289°C to 291°C
Quantity 25 g
Beilstein 3703311
Merck Index 14,9802
Formula Weight 328.51
Percent Purity 99%
Chemical Name or Material (+/-)-2-Aminoheptane sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.