missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoheptane, MedChemExpress

Product Code. 16472289
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16472289 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16472289 Supplier Medchem Express Supplier No. HYB0952
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 2-Aminoheptane (1-Methylhexylamine) is an isomeric heptylamine commonly used as stimulant. 2-Aminoheptane can be used as the nitrogen source in minimal medium.

2-Aminoheptane (1-Methylhexylamine) is an isomeric heptylamine commonly used as stimulant. 2-Aminoheptane can be used as the nitrogen source in minimal medium.

Specifications

Chemical Name or Material 2-Aminoheptane
Physical Form Liquid
CAS 123-82-0
Color Colorless
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C7H17N
Synonym 1-Methylhexylamine 2-Heptylamine
Solubility Information DMSO : ≥ 100 mg/mL (867.90 mM)
SMILES CC(N)CCCCC
Molecular Weight (g/mol) 115.22
Formula Weight 115.22
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.