missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoethyl Sulfate, TRC

Product Code. 30327512
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327512 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327512 Supplier Toronto Research Chemicals Supplier No. TRCA60954550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Aminoethyl Sulfate
CAS 926-39-6
Molecular Formula C2H7NO4S
Quantity 50 g
Synonym 2-Aminoethanol Hydrogen Sulfate (Ester), 2-Aminoethyl Hydrogen Sulfate, 2-Aminoethyl Sulfate, 2-Aminoethyl Sulfuric Acid, Ethanolamine O-Sulfate, Ethanolamine Sulfate, Mono(2-aminoethyl) Sulfate, NSC 204188, NSC 3532, WAS-34
SMILES NCCOS(=O)(=O)O
Molecular Weight (g/mol) 141.15
Formula Weight 141.0096
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.