missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoethyl hydrogen sulfate, 99%

Product Code. 10539790 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10539790 500 g Plastic bottle 500g
10375780 100 g Plastic bottle 100g
10493205 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10539790 Supplier Thermo Scientific Acros Supplier No. 103645000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 926-39-6 | C2H7NO4S | 141.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 926-39-6
Molecular Formula C2H7NO4S
Molecular Weight (g/mol) 141.14
MDL Number MFCD00008179
InChI Key WSYUEVRAMDSJKL-UHFFFAOYSA-N
Synonym ethanolamine o-sulfate, aminoethyl sulfate, 2-aminoethyl sulfate, mono 2-aminoethyl sulfate, 2-aminoethyl hydrogen sulphate, sulfuric acid mono 2-aminoethyl ester, was-34, 2-aminoethylhydrogensulfate, unii-9c8f910hlk, ethanol, 2-amino-, hydrogen sulfate ester
PubChem CID 70223
IUPAC Name 2-aminoethyl hydrogen sulfate
SMILES NCCOS(O)(=O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 277°C to 280°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula H2NCH2CH2OSO3H
Quantity 500 g
Beilstein 04, 276
Solubility Information Solubility in water: soluble. Other solubilities: insolube in ethanol
Formula Weight 141.14
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Aminoethyl hydrogen sulfate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.