missing translation for 'onlineSavingsMsg'
Learn More

(2-Aminoethoxy)acetic acid, 98%

Product Code. 15458907
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15458907 1 g 1g
15468907 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15458907 Supplier Thermo Scientific Alfa Aesar Supplier No. H52289.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 10366-71-9 | C4H9NO3 | 119.12 g/mol

Reactant in the synthesis ammonium ion-selective fluoroionophore. Used in the analysis of recognition elements for mitochondrial processing peptidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the synthesis ammonium ion-selective fluoroionophore. Used in the analysis of recognition elements for mitochondrial processing peptidase.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10366-71-9
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD09865417
InChI Key GNRLUBOJIGSVNT-UHFFFAOYSA-N
Synonym 2-2-aminoethoxy acetic acid, 2-amino-ethoxy-acetic acid, 2-aminoethoxy acetic acid, acetic acid, aminoethoxy, 5-amino-3-oxapentanoic acid, aminoethoxyacetic acid, 2-2-aminoethoxyl acetic acid, acetic acid, 2-aminoethoxy
PubChem CID 11170956
IUPAC Name 2-(2-aminoethoxy)acetic acid
SMILES C(COCC(=O)O)N

Specifications

Melting Point 177°C to 179°C
Linear Formula NH2CH2CH2OCH2CO2H
Quantity 1 g
Formula Weight 119.12
Percent Purity 98%
Chemical Name or Material (2-Aminoethoxy)acetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.