missing translation for 'onlineSavingsMsg'
Learn More

[2-(Aminocarbonyl)phenoxy]acetic Acid, TRC

Product Code. 30324818
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30324818 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324818 Supplier Toronto Research Chemicals Supplier No. TRCA615628250MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material [2-(Aminocarbonyl)phenoxy]acetic Acid
CAS 25395-22-6
Molecular Formula C9H9NO4
Quantity 250 mg
Synonym 2-[2-(Aminocarbonyl)phenoxy]acetic Acid, (2-Carbamoylphenoxy)acetic Acid, (o-Carbamoylphenoxy)acetic Acid, 2-(2-Carbamoylphenoxy)acetic Acid, 2-(2-Carbamoylphenoxy)aceticacid, 2-(Carboxymethoxy)benzamide, NSC 65589Salicylamide O-Acetic Acid, [2-(Aminocarbonyl)phenoxy]acetic Acid, o-Carboxyamidophenoxyacetic Acid
InChI Formula InChI=1S/C9H9NO4/c10-9(13)6-3-1-2-4-7(6)14-5-8(11)12/h1-4H,5H2,(H2,10,13)(H,11,12)
SMILES NC(=O)c1ccccc1OCC(=O)O
IUPAC Name 2-(2-carbamoylphenoxy)acetic acid
Molecular Weight (g/mol) 195.17
Formula Weight 195.0532
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.