missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzyl alcohol, 98%

Product Code. 11494574
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494574 250 g 250g
11474574 10 g 10g
11484574 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494574 Supplier Thermo Scientific Alfa Aesar Supplier No. A13180.30
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5344-90-1 | C7H9NO | 123.155 g/mol

2-Aminobenzyl alcohol is oxidatively cyclised with an array of ketones in dioxane at 80°C in the presence of a ruthenium catalyst and KOH to give corresponding quinolines. It undergoes oxidation catalyzed by heterotrimetallic RuMnMn species on the hydrotalcite surface in the presence of O2 to yield 2-aminobenzaldehyde. was used in the synthesis of ethyl 2-hydroxymethylcarbanilate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminobenzyl alcohol is oxidatively cyclised with an array of ketones in dioxane at 80°C in the presence of a ruthenium catalyst and KOH to give corresponding quinolines. It undergoes oxidation catalyzed by heterotrimetallic RuMnMn species on the hydrotalcite surface in the presence of O2 to yield 2-aminobenzaldehyde. was used in the synthesis of ethyl 2-hydroxymethylcarbanilate.

Solubility
Very soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5344-90-1
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00007749
InChI Key VYFOAVADNIHPTR-UHFFFAOYSA-N
Synonym 2-aminobenzyl alcohol, 2-aminophenyl methanol, 2-aminobenzylalcohol, o-aminobenzyl alcohol, 2-aminobenzenemethanol, benzenemethanol, 2-amino, o-aminobenzylic alcohol, o-hydroxymethyl aniline, benzyl alcohol, o-amino, 2-hydroxymethylaniline
PubChem CID 21439
IUPAC Name (2-aminophenyl)methanol
SMILES C1=CC=C(C(=C1)CO)N

Specifications

Melting Point 81°C to 84°C
Boiling Point 160°C (10 mmHg)
Quantity 250 g
Beilstein 1072211
Sensitivity Air and light sensitive
Solubility Information Very soluble in water.
Formula Weight 123.16
Percent Purity 98%
Chemical Name or Material 2-Aminobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.