missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzotrifluoride, 99%, Thermo Scientific Chemicals

Catalog Number p-55269
Click to view available options
Quantity:
5 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
88-17-5
C7H6F3N
161.13
MFCD00007718
VBLXCTYLWZJBKA-UHFFFAOYSA-N
2-trifluoromethyl aniline, 2-aminobenzotrifluoride, o-aminobenzotrifluoride, o-trifluoromethyl aniline, o-trifluoromethylaniline, benzenamine, 2-trifluoromethyl, 2-trifluoromethyl benzenamine, alpha,alpha,alpha-trifluoro-o-toluidine, 2-aminotrifluorotoluene, 2-aminotrifluoromethylbenzene
6922
2-(trifluoromethyl)aniline
C1=CC=C(C(=C1)C(F)(F)F)N
This item is not returnable. View return policy
88-17-5
C7H6F3N
161.13
MFCD00007718
VBLXCTYLWZJBKA-UHFFFAOYSA-N
2-trifluoromethyl aniline, 2-aminobenzotrifluoride, o-aminobenzotrifluoride, o-trifluoromethyl aniline, o-trifluoromethylaniline, benzenamine, 2-trifluoromethyl, 2-trifluoromethyl benzenamine, alpha,alpha,alpha-trifluoro-o-toluidine, 2-aminotrifluorotoluene, 2-aminotrifluoromethylbenzene
6922
2-(trifluoromethyl)aniline
C1=CC=C(C(=C1)C(F)(F)F)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -33.0°C
Color Amber to Colorless
Density 1.2960g/mL
Boiling Point 170.0°C to 173.0°C (15.0 mmHg)
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H4CF3
Refractive Index 1.4800 to 1.4820
Quantity 5 mL
Beilstein 12, II, 453
Specific Gravity 1.296
Solubility Information Solubility in water: 4g/L in water (20°C). Other solubilities: soluble in ether,ethanol and diethyl ether
Formula Weight 161.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Aminobenzotrifluoride, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Aminobenzotrifluoride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'