missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminobenzothiazole, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 1,3-Benzothiazol-2-amine |
| CAS | 136-95-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C7 H6 N2 S |
| Quantity | 10 g |
| Synonym | 2-Benzothiazolamine, Benzothiazole, 1-amino- (3CI), Benzothiazole, 2-amino- (6CI,7CI,8CI), Benzothiazoline, 2-imino- (4CI), 1,3-Benzothiazol-2-amine, 1,3-Benzothiazol-2-ylamine, 2(3H)-Benzothiazolimine, 2-Aminobenzo[d]thiazole, 2-Aminobenzothiazole, 2-Benzothiazolylamine, 2-Iminobenzothiazoline, ABT, Benzo[d]thiazol-2-amine, Cerium ionophore I, NSC 4670, SKA 1 |
| InChI Formula | InChI=1S/C7H6N2S/c8-7-9-5-3-1-2-4-6(5)10-7/h1-4H,(H2,8,9) |
| SMILES | Nc1nc2ccccc2s1 |
| IUPAC Name | 1,3-benzothiazol-2-amine |
| Molecular Weight (g/mol) | 150.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction