missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzothiazole, 97%

Product Code. 10635582 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10635582 5 g Glass bottle 5g
10593881 100 g Plastic bottle 100g
10503891 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10635582 Supplier Thermo Scientific Acros Supplier No. 148810050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 136-95-8 | C7H6N2S | 150.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136-95-8
Molecular Formula C7H6N2S
Molecular Weight (g/mol) 150.2
MDL Number MFCD00005785
InChI Key UHGULLIUJBCTEF-UHFFFAOYSA-N
Synonym 2-aminobenzothiazole, 2-benzothiazolamine, benzo d thiazol-2-amine, benzothiazol-2-ylamine, 2-aminobenzthiazole, benzothiazole-2-ylamine, 2-iminobenzothiazoline, o-aminobenzothiazole, benzothiazole, 2-amino, 2-benzothiazolylamine
PubChem CID 8706
IUPAC Name 1,3-benzothiazol-2-amine
SMILES C1=CC=C2C(=C1)N=C(S2)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 129°C to 134°C
Color Beige to Gray
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Beilstein 27, 182
Merck Index 15, 419
Solubility Information Solubility in water: sparingly soluble in water. Other solubilities: soluble in concentrated acids,freely soluble in alcohol,ether and chloroform
Formula Weight 150.2
Percent Purity 97%
Physical Form Powder or Flakes
Chemical Name or Material 2-Aminobenzothiazole, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.