missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzophenone, TRC

Product Code. 30324253
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30324253 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324253 Supplier Toronto Research Chemicals Supplier No. TRCA625780100G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Aminobenzophenone
CAS 2835-77-0
Molecular Formula C13H11NO
Quantity 100 g
Synonym (2-Aminophenyl)phenylmethanone, 2-(Phenylcarbonyl)aniline, 2-Aminobenzophenone, 2-Aminophenyl Phenyl Ketone, 2-Benzoylaniline, 2-Benzoylbenzenamine, NSC 9422, o-Aminobenzophenone, o-Aminophenyl Phenyl Ketone, o-Benzoylaniline
InChI Formula InChI=1S/C13H11NO/c14-12-9-5-4-8-11(12)13(15)10-6-2-1-3-7-10/h1-9H,14H2
SMILES Nc1ccccc1C(=O)c2ccccc2
IUPAC Name (2-aminophenyl)-phenylmethanone
Molecular Weight (g/mol) 197.23
Formula Weight 197.0841
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.