missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzonitrile, 98%

Product Code. 10162320 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10162320 10 g Glass bottle 10g
10539630 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10162320 Supplier Thermo Scientific Acros Supplier No. 104920100
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1885-29-6 | C7H6N2 | 118.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1885-29-6
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00007631
InChI Key HLCPWBZNUKCSBN-UHFFFAOYSA-N
Synonym anthranilonitrile, 2-cyanoaniline, o-aminobenzonitrile, benzonitrile, 2-amino, o-cyanoaniline, 2-amino-benzonitrile, aminobenzonitrile, aminobenzonitrile 2-, 2-aminobenzenecarbonitrile, unii-5rxh2b211r
PubChem CID 72913
IUPAC Name 2-aminobenzonitrile
SMILES NC1=CC=CC=C1C#N

Specifications

Melting Point 46.0°C to 50.0°C
Boiling Point 267.0°C to 268.0°C
Flash Point 145°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H4CN
Quantity 10 g
Beilstein 14, 322
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 118.14
Percent Purity 98%
Physical Form Crystalline Flakes
Chemical Name or Material 2-Aminobenzonitrile, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Anthranilonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.