missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzimidazole, Mikromol™

Product Code. 17980606
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17980606 100 mg 100mg
17911406 25 mg 25mg
30463860 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17980606 Supplier Mikromol Supplier No. MM2500.03
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-Aminobenzimidazole
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Aminobenzimidazole
CAS 934-32-7
Analyte or Component Names 2-Aminobenzimidazole
Shipping Condition Room Temperature
Molecular Formula C7 H7 N3
Certifications/Compliance ISO 17025
Synonym 2-Aminobenzimidazole, 1H-Benzimidazol-2-amine, Benzimidazole, 2-amino- (6CI,7CI,8CI), 1H-Benzimidazol-2-ylamine, 2-AB, 2-Amino-1H-benzimidazole, 2-Aminobenzimidazole, 2-Benzimidazolamine, 2-Benzimidazolylamine, 2-Iminobenzimidazoline, NSC 27793, NSC 7628
InChI Formula InChI=1S/C7H7N3/c8-7-9-5-3-1-2-4-6(5)10-7/h1-4H,(H3,8,9,10)
SMILES Nc1nc2ccccc2[nH]1
IUPAC Name 1H-benzimidazol-2-amine
Molecular Weight (g/mol) 133.15
Quantity 100 mg
Formula Weight 133.064 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.